Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amigo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164563.1 | NP_001158035.1 | 2 | 1515 | ||
NM_001164602.1 | NP_001158074.1 | 2 | 1483 | |||
NM_178114.4 | NP_835215.1 | 1 | 1858 | |||
GenBank mRNA | AB078880.1 | - | 1 | 1516 | ||
AK030848.1 | - | 1 | 1825 | |||
AK038783.1 | - | 2 | 1517 | |||
AY237006.1 | - | 1 | 1110 | |||
BC095990.1 | - | 1 | 1693 | 5358186 |