Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPCML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012393.2 | NP_001012393.1 | 3 | 655 | ||
NM_001319103.1 | NP_001306032.1 | 2 | 933 | |||
NM_001319104.1 | NP_001306033.1 | 2 | 570 | |||
NM_001319105.1 | NP_001306034.1 | 3 | 479 | |||
NM_001319106.1 | NP_001306035.1 | 2 | 933 | |||
NM_002545.4 | NP_002536.1 | 2 | 933 | |||
XM_006718846.2 | XP_006718909.1 | 3 | 881 | |||
XM_011542856.2 | XP_011541158.1 | 4 | 1191 | |||
GenBank mRNA | AK289695.1 | - | 2 | 561 | ||
AK299528.1 | - | 4 | 328 | |||
AK299908.1 | - | 3 | 442 | |||
AK314077.1 | - | 2 | 933 | |||
AK315859.1 | - | 2 | 260 | |||
AK316273.1 | - | 2 | 357 | |||
AK316491.1 | - | 3 | 479 | |||
BC074742.2 | - | 2 | 415 | 30915458 | ||
BC074773.2 | - | 2 | 415 | 30915200 | ||
BC117254.1 | - | 2 | 411 | 40125805 | ||
BC126251.1 | - | 2 | 411 | 8991967 | ||
BC143945.1 | - | 2 | 411 | 9052463 | ||
BC143946.1 | - | 2 | 411 | 9052464 | ||
BX537377.1 | - | 3 | 503 | |||
EU562295.1 | - | 2 | 967 | |||
EU562296.1 | - | 3 | 445 | |||
EU562297.1 | - | 3 | 382 | |||
EU562298.1 | - | 2 | 360 | |||
EU562299.1 | - | 2 | 272 | |||
EU562300.1 | - | 1 | 196 | |||
L34774.1 | - | 2 | 438 |