Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPCML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012393.2 | NP_001012393.1 | 5, 6 | 938 | ||
NM_001319103.1 | NP_001306032.1 | 4, 5 | 1216 | |||
NM_001319104.1 | NP_001306033.1 | 4, 5 | 853 | |||
NM_001319105.1 | NP_001306034.1 | 5, 6 | 762 | |||
NM_001319106.1 | NP_001306035.1 | 4, 5 | 1213 | |||
NM_002545.4 | NP_002536.1 | 4, 5 | 1216 | |||
XM_006718846.2 | XP_006718909.1 | 5, 6 | 1161 | |||
XM_011542856.2 | XP_011541158.1 | 6, 7 | 1474 | |||
GenBank mRNA | AK289695.1 | - | 4, 5 | 844 | ||
AK299528.1 | - | 6, 7 | 611 | |||
AK299908.1 | - | 5, 6 | 725 | |||
AK314077.1 | - | 4, 5 | 1213 | |||
AK315859.1 | - | 4, 5 | 543 | |||
AK316273.1 | - | 4, 5 | 640 | |||
AK316491.1 | - | 5, 6 | 762 | |||
BC074742.2 | - | 4, 5 | 695 | 30915458 | ||
BC074773.2 | - | 4, 5 | 698 | 30915200 | ||
BC117254.1 | - | 4, 5 | 694 | 40125805 | ||
BC126251.1 | - | 4, 5 | 694 | 8991967 | ||
BC143945.1 | - | 4, 5 | 691 | 9052463 | ||
BC143946.1 | - | 4, 5 | 694 | 9052464 | ||
BX537377.1 | - | 5, 6 | 786 | |||
EU562295.1 | - | 4, 5 | 1250 | |||
EU562296.1 | - | 5, 6 | 728 | |||
EU562297.1 | - | 5, 6 | 665 | |||
EU562298.1 | - | 4, 5 | 643 | |||
EU562299.1 | - | 4, 5 | 555 | |||
EU562300.1 | - | 3, 4 | 479 | |||
L34774.1 | - | 4, 5 | 721 |