Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178919.4 | NP_849250.1 | 9, 10 | 1466 | ||
GenBank mRNA | AK042427.1 | - | 9, 10 | 1426 | ||
AK084606.1 | - | 9, 10 | 1292 | |||
AK149916.1 | - | 9, 10 | 1418 | |||
AK149934.1 | - | 9, 10 | 1467 | |||
AK158371.1 | - | 8, 9 | 1521 | |||
AK171488.1 | - | 9, 10 | 1447 | |||
AK188756.1 | - | 1, 2 | 64 | |||
BC026448.1 | - | 8, 9 | 1246 | |||
BC038039.1 | - | 8, 9 | 1968 | |||
BC079532.1 | - | 9, 10 | 1362 | 6403184 |