Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178920.4 | NP_849251.1 | 4 | 618 | ||
GenBank mRNA | AK035986.1 | - | 4 | 500 | ||
BC021848.1 | - | 1 | 29 | |||
BC113154.1 | - | 4 | 495 | 40058489 |