Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207216.2 | NP_997099.2 | 3 | 409 | ||
GenBank mRNA | AI746748.1 | - | 3 | 388 | ||
AK128903.1 | - | 3 | 381 | |||
AK143745.1 | - | 3 | 387 | |||
BC025940.1 | - | 3 | 376 | 5100753 |