Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc45a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033219.3 | NP_001028391.2 | 5 | 1488 | ||
NM_001168255.1 | NP_001161727.1 | 4 | 566 | |||
GenBank mRNA | AK133269.1 | - | 5 | 1489 | ||
AK170738.1 | - | 4 | 571 | |||
BC056501.1 | - | 5 | 1085 | 5720940 | ||
BC058722.1 | - | 4 | 484 |