Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cggbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178647.2 | NP_848762.1 | 3 | 455 | ||
GenBank mRNA | AK049631.1 | - | 3 | 533 | ||
AK052195.1 | - | 3 | 523 | |||
AK083450.1 | - | 3 | 460 | |||
BC043097.1 | - | 3 | 455 | 6405954 | ||
BC057053.1 | - | 3 | 509 | 6827183 |