Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPHN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002547.2 | NP_002538.1 | 3, 4 | 551 | ||
XM_005262270.1 | XP_005262327.1 | 3, 4 | 621 | |||
XM_006724653.2 | XP_006724716.1 | 3, 4 | 621 | |||
XM_011530961.1 | XP_011529263.1 | 3, 4 | 472 | |||
XM_017029555.1 | XP_016885044.1 | 3, 4 | 621 | |||
GenBank mRNA | AB102656.1 | - | 2, 3 | 257 | ||
AJ001189.1 | - | 3, 4 | 895 | |||
AK295769.1 | - | 3, 4 | 621 | |||
BC059393.1 | - | 3, 4 | 551 | |||
BC140763.1 | - | 3, 4 | 589 | 9021631 | ||
BP357826.1 | - | 3, 4 | 551 |