Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC027330.1 | - | 23 | 2924 | 3674968 | |
BC029320.1 | - | 23 | 2925 |