Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039521.1 | NP_001034610.1 | 11, 12 | 1056 | ||
GenBank mRNA | AK080397.1 | - | 11, 12 | 1012 | ||
AK080680.1 | - | 1037 | ||||
AK165676.1 | - | 11, 12 | 1045 | |||
BC034110.1 | - | 7, 8 | 571 | |||
BC138774.1 | - | 11, 12 | 1021 | 8861796 | ||
BC138776.1 | - | 11, 12 | 1021 | 8861798 |