Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145480.1 | NP_663455.1 | 7, 8 | 733 | ||
GenBank mRNA | AK146954.1 | - | 7, 8 | 721 | ||
AK147030.1 | - | 7, 8 | 721 | |||
BC003335.1 | - | 7, 8 | 733 | 3257208 |