Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc51a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145932.3 | NP_666044.1 | 4 | 697 | ||
GenBank mRNA | AK052725.1 | - | 4 | 698 | ||
AK147155.1 | - | 517 | ||||
BC024441.1 | - | 4 | 505 | 4973857 | ||
BC025912.1 | - | 4 | 508 | 5064489 | ||
BC031178.1 | - | 4 | 448 | 4988213 |