Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkdcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134117.2 | NP_598878.2 | 3 | 1036 | ||
GenBank mRNA | AB381893.1 | - | 3 | 1028 | ||
AB436780.1 | - | 3 | 1017 | |||
BC022157.1 | - | 2 | 183 | |||
BC086639.1 | - | 4 | 874 | 6828629 |