Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134123.3 | NP_598884.2 | 20 | 3788 | ||
GenBank mRNA | AK032418.1 | - | 1990 | |||
AK088735.1 | - | 1 | 1024 | |||
AK122451.1 | - | 20 | 3655 | |||
AK161113.1 | - | 6 | 1700 | |||
BC002302.1 | - | 1 | 607 | |||
BC030846.1 | - | 7 | 1786 | |||
BC038363.1 | - | 20 | 3432 | 5371946 | ||
BC075621.1 | - | 20 | 3781 | 6406441 |
Gene Symbol: C530047H08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC040266.1 | - | 506 |