Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134123.3 | NP_598884.2 | 20 | 3998 | ||
GenBank mRNA | AK032418.1 | - | 2200 | |||
AK088735.1 | - | 1 | 1234 | |||
AK122451.1 | - | 20 | 3865 | |||
AK161113.1 | - | 6 | 1910 | |||
BC002302.1 | - | 1 | 817 | |||
BC030846.1 | - | 7 | 1996 | |||
BC038363.1 | - | 20 | 3642 | 5371946 | ||
BC075621.1 | - | 20 | 3991 | 6406441 |
Gene Symbol: C530047H08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC040266.1 | - | 716 |