Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025572.1 | NP_001020743.1 | 5 | 594 | ||
GenBank mRNA | AK012645.1 | - | 4 | 518 | ||
AK013457.1 | - | 4 | 569 | |||
AK133715.1 | - | 5 | 593 | |||
AK153850.1 | - | 4 | 503 | |||
BC023046.1 | - | 5 | 465 | |||
BC050185.1 | - | 5 | 574 | |||
BC064777.1 | - | 4 | 509 | |||
BC080825.1 | - | 4 | 517 | |||
BC094292.1 | - | 4 | 515 |