Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025572.1 | NP_001020743.1 | 6 | 820 | ||
GenBank mRNA | AK133715.1 | - | 6 | 819 | ||
AK153850.1 | - | 5 | 729 | |||
BC023046.1 | - | 6 | 691 | |||
BC050185.1 | - | 6 | 800 | |||
BC064777.1 | - | 5 | 735 | |||
BC080825.1 | - | 5 | 743 |