Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oard1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289490.1 | NP_001276419.1 | 3, 4 | 588 | ||
NM_001289491.1 | NP_001276420.1 | 3, 4 | 313 | |||
NM_207219.4 | NP_997102.2 | 3, 4 | 453 | |||
GenBank mRNA | AK148136.1 | - | 3, 4 | 575 | ||
AK153512.1 | - | 3, 4 | 376 | |||
BC022574.1 | - | 2, 3 | 232 | 3989373 | ||
BC157987.1 | - | 3, 4 | 338 | 9007303 |