Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgxb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134134.2 | NP_598895.2 | 17 | 3137 | ||
NM_178277.1 | NP_840061.1 | 16 | 3232 | |||
GenBank mRNA | AB045322.1 | - | 17 | 3138 | ||
AK129085.1 | - | 13 | 2140 | |||
AK132529.1 | - | 10 | 1616 | |||
BC025148.1 | - | 2 | 100 | |||
BC058089.1 | - | 14 | 2530 | |||
BC060046.1 | - | 10 | 1562 | |||
BC066029.1 | - | 11 | 2368 | 6417061 | ||
BC079580.1 | - | 16 | 3180 | 5702202 | ||
BG061435.2 | - | 2 | 228 |