Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgxb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134134.2 | NP_598895.2 | 4 | 859 | ||
NM_178277.1 | NP_840061.1 | 4 | 858 | |||
GenBank mRNA | AB045322.1 | - | 4 | 859 | ||
AK041866.1 | - | 4 | 837 | |||
AK144993.1 | - | 4 | 843 | |||
BC058089.1 | - | 2 | 156 | |||
BC079580.1 | - | 4 | 806 | 5702202 |