Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205336.1 | NP_001192265.1 | 20 | 3066 | ||
NM_139206.2 | NP_631945.2 | 20 | 2866 | |||
GenBank mRNA | AF469621.1 | - | 19 | 2816 | ||
AF469622.1 | - | 20 | 3007 | |||
AK220486.1 | - | 19 | 3295 | |||
BC068145.1 | - | 20 | 2854 | 6856630 |