Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Me2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145494.2 | NP_663469.1 | 11 | 1177 | ||
GenBank mRNA | AK042289.1 | - | 11 | 1224 | ||
AK043103.1 | - | 11 | 1214 | |||
AK050933.1 | - | 11 | 1224 | |||
AK050980.1 | - | 11 | 1224 | |||
AK077746.1 | - | 11 | 1224 | |||
AK156403.1 | - | 11 | 1224 | |||
AK162126.1 | - | 11 | 1236 | |||
AK171157.1 | - | 11 | 1224 | |||
BC004709.1 | - | 11 | 1177 | 3499365 |