Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167875.1 | NP_001161347.1 | 4 | 636 | ||
NM_001167877.1 | NP_001161349.1 | 4 | 636 | |||
NM_134144.2 | NP_598905.2 | 4 | 636 | |||
GenBank mRNA | AK149466.1 | - | 4 | 581 | ||
AY206873.1 | - | 4 | 615 | |||
BC011222.1 | - | 4 | 619 | 4195223 | ||
BC051050.1 | - | 4 | 627 | 5052536 |