Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134150.2 | NP_598911.1 | 5, 6 | 452 | ||
GenBank mRNA | AK027996.1 | - | 5, 6 | 439 | ||
AK087195.1 | - | 5, 6 | 439 | |||
AK140070.1 | - | 5, 6 | 439 | |||
AK145273.1 | - | 5, 6 | 442 | |||
AK145970.1 | - | 5, 6 | 444 | |||
BC022575.1 | - | 5, 6 | 452 | 5005025 | ||
BC054410.1 | - | 5, 6 | 420 |