Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps37c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181403.2 | NP_852068.1 | 4 | 502 | ||
GenBank mRNA | AK030717.1 | - | 3 | 373 | ||
AK150606.1 | - | 4 | 554 | |||
AK158833.1 | - | 3 | 241 | |||
AK159309.1 | - | 4 | 575 | |||
AK165826.1 | - | 4 | 557 | |||
AK189154.1 | - | 2 | 94 | |||
BC025865.1 | - | 4 | 502 | 5029143 |