Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178930.3 | NP_849261.2 | 23, 24 | 3216 | ||
GenBank mRNA | AK172919.1 | - | 20, 21 | 2809 | ||
BC024129.1 | - | 6, 7 | 874 | |||
BC034745.1 | - | 8, 9 | 981 | |||
BC060728.1 | - | 15, 16 | 2636 | |||
BC076569.1 | - | 23, 24 | 3216 | 6409170 | ||
BC082336.1 | - | 24, 25 | 3211 |