Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178930.3 | NP_849261.2 | 22 | 3015 | ||
GenBank mRNA | AK161861.1 | - | 1 | 158 | ||
AK172919.1 | - | 19 | 2608 | |||
BC024129.1 | - | 5 | 673 | |||
BC034745.1 | - | 7 | 780 | |||
BC060728.1 | - | 14 | 2435 | |||
BC076569.1 | - | 22 | 3015 | 6409170 | ||
BC082336.1 | - | 23 | 3010 |