Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012368.2 | NP_036500.2 | 1 | 171 | ||
GenBank mRNA | BC069158.1 | - | 1 | 119 | 7217019 | |
BC069456.1 | - | 1 | 119 | 7262277 | ||
BC126269.1 | - | 1 | 171 | 8991985 | ||
BC130328.1 | - | 1 | 171 | 40146359 |