Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030693.2 | NP_109618.1 | 3 | 1104 | ||
GenBank mRNA | AB012276.1 | - | 1 | 214 | ||
AF310624.1 | - | 3 | 1125 | |||
AF312938.1 | - | 3 | 1080 | |||
AF375475.1 | - | 3 | 1109 | |||
AF375476.1 | - | 3 | 1104 | |||
AK131939.1 | - | 3 | 1126 | |||
AK146479.1 | - | 3 | 1126 | |||
AK159624.1 | - | 3 | 1128 | |||
AK160085.1 | - | 1 | 529 | |||
AY024362.1 | - | 1 | 619 | |||
BC010195.1 | - | 2 | 937 | 3157499 | ||
BC092068.1 | - | 3 | 1109 | 4210931 |