Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172671.2 | NP_766259.2 | 11, 12 | 1526 | ||
GenBank mRNA | AK044357.1 | - | 10, 11 | 834 | ||
CF164628.1 | - | 7, 8 | 548 | |||
CN667965.1 | - | 2, 3 | 98 |