Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252374.1 | NP_001239303.1 | 4 | 221 | ||
NM_026004.3 | NP_080280.3 | 3 | 378 | |||
GenBank mRNA | AK011525.1 | - | 3 | 329 | ||
AK011894.1 | - | 3 | 276 | |||
AK160139.1 | - | 4 | 221 | |||
AK208033.1 | - | 3 | 141 | |||
BC038029.1 | - | 4 | 214 | 4018956 | ||
CB571852.1 | - | 4 | 226 |