Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col16a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028266.5 | NP_082542.3 | 68, 69 | 4747 | ||
GenBank mRNA | AK012212.1 | - | 1, 2 | 379 | ||
AK031163.1 | - | 59, 60 | 3612 | |||
AK040971.1 | - | 68, 69 | 4700 | |||
BC023940.1 | - | 22, 23 | 1456 | |||
BC027766.1 | - | 68, 69 | 4682 | 5346676 |