Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171007.1 | NP_001164478.1 | 4 | 2406 | ||
NM_172729.3 | NP_766317.1 | 4 | 2657 | |||
GenBank mRNA | AK082663.1 | - | 1 | 1452 | ||
AK089662.1 | - | 4 | 2407 | |||
AK137585.1 | - | 4 | 2662 | |||
AY160222.1 | - | 3 | 2192 | |||
BC042670.1 | - | 4 | 2484 | 3597911 | ||
BC043670.1 | - | 1 | 1374 |