Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039086.1 | NP_001034175.1 | 26 | 3894 | ||
NM_001039087.1 | NP_001034176.1 | 26 | 3912 | |||
NM_054050.2 | NP_473391.1 | 24 | 3498 | |||
GenBank mRNA | AF348669.1 | - | 24 | 3454 | ||
AK077034.1 | - | 16 | 1884 | |||
AK147468.1 | - | 26 | 3913 | |||
AK147470.1 | - | 26 | 3913 | |||
AK147506.1 | - | 26 | 3881 | |||
AK147696.1 | - | 26 | 3874 | |||
BC034081.1 | - | 7 | 663 | |||
BC039756.1 | - | 12 | 1225 | |||
BC129881.1 | - | 25 | 3605 | 40110588 |