Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039086.1 | NP_001034175.1 | 6 | 1046 | ||
NM_001039087.1 | NP_001034176.1 | 7 | 1160 | |||
NM_054050.2 | NP_473391.1 | 7 | 1160 | |||
GenBank mRNA | AF348669.1 | - | 7 | 1116 | ||
AK147468.1 | - | 7 | 1161 | |||
AK147470.1 | - | 7 | 1161 | |||
AK147506.1 | - | 7 | 1129 | |||
AK147696.1 | - | 6 | 1026 | |||
BC129881.1 | - | 6 | 856 | 40110588 |