Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006190.4 | NP_006181.1 | 13 | 1393 | ||
NR_033915.1 | 13 | 1393 | ||||
XM_006712555.3 | XP_006712618.1 | 13 | 1340 | |||
XM_011511252.2 | XP_011509554.1 | 14 | 1455 | |||
XM_011511253.2 | XP_011509555.1 | 14 | 1327 | |||
GenBank mRNA | AK302614.1 | - | 13 | 1365 | ||
AK316433.1 | - | 13 | 1294 | |||
BC014834.1 | - | 13 | 1336 | 2899929 | ||
U27459.1 | - | 13 | 1327 | |||
U40268.2 | - | 13 | 1308 |