Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm6sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145375.4 | NP_663350.2 | 1, 2 | 543 | ||
GenBank mRNA | AK046714.1 | - | 1, 2 | 444 | ||
AK083132.1 | - | 1, 2 | 546 | |||
AK135100.1 | - | 1, 2 | 525 | |||
AK162572.1 | - | 1, 2 | 395 | |||
AK163621.1 | - | 1, 2 | 468 | |||
AK172109.1 | - | 1, 2 | 459 | |||
AK172145.1 | - | 1, 2 | 444 | |||
BB653227.1 | - | 1, 2 | 545 | |||
BC023123.1 | - | 1, 2 | 425 | 3995280 |