Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmjd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033398.2 | NP_203971.2 | 3 | 905 | ||
GenBank mRNA | AF304118.1 | - | 3 | 838 | ||
AK122317.1 | - | 3 | 746 | |||
BC006067.1 | - | 3 | 714 | |||
BC011503.1 | - | 3 | 737 | |||
BC056629.1 | - | 3 | 905 | 5033487 | ||
BC117737.1 | - | 2 | 546 | |||
CX210791.1 | - | 3 | 611 | |||
EF527404.1 | - | 3 | 905 | |||
EF527405.1 | - | 3 | 691 |