Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thoc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172438.3 | NP_766026.1 | 20 | 2064 | ||
GenBank mRNA | AK006840.1 | - | 11 | 1071 | ||
AK050734.1 | - | 20 | 2064 | |||
AK173078.1 | - | 18 | 1902 | |||
BC020332.1 | - | 19 | 2029 | |||
BC021779.1 | - | 19 | 2037 | |||
BC039758.1 | - | 20 | 2065 | 3995737 |