Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174991.3 | NP_778156.2 | 5 | 2050 | ||
GenBank mRNA | AK147456.1 | - | 5 | 2035 | ||
AK147459.1 | - | 5 | 2003 | |||
AK147494.1 | - | 5 | 1837 | |||
AK147600.1 | - | 5 | 1812 | |||
AK147607.1 | - | 5 | 2054 | |||
AK220481.1 | - | 5 | 1429 | |||
AY168408.1 | - | 5 | 1562 |
Set Membership: |
G-protein coupled receptor |