Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008104.2 | NP_032130.2 | 5 | 1003 | ||
GenBank mRNA | AF081556.1 | - | 5 | 969 | ||
AK132499.1 | - | 3 | 787 | |||
AK135689.1 | - | 7 | 1002 | |||
BC110631.1 | - | 5 | 1033 | 40054293 | ||
BC110632.1 | - | 5 | 1033 | 40054295 | ||
D88611.1 | - | 5 | 1210 |