Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145979.2 | NP_666091.1 | 8 | 1097 | ||
GenBank mRNA | AK034549.1 | - | 3 | 178 | ||
AK145532.1 | - | 8 | 1084 | |||
AK147695.1 | - | 8 | 1280 | |||
AK220473.1 | - | 7 | 1005 | |||
BC058578.1 | - | 8 | 1097 | 6489649 |