Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145979.2 | NP_666091.1 | 22 | 3432 | ||
GenBank mRNA | AK034549.1 | - | 17 | 2513 | ||
AK147695.1 | - | 22 | 3576 | |||
AK153826.1 | - | 15 | 2190 | |||
AK220473.1 | - | 21 | 3340 | |||
BC058578.1 | - | 22 | 3432 | 6489649 | ||
BC059082.1 | - | 9 | 1213 |