Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pom121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148932.2 | NP_683734.2 | 11, 12 | 3428 | ||
GenBank mRNA | AF516680.1 | - | 11, 12 | 3389 | ||
AK154473.1 | - | 11, 12 | 3429 | |||
AK172982.1 | - | 10, 11 | 2807 | |||
BC053101.1 | - | 11, 12 | 3404 | 6405486 |