Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130860.3 | NP_570930.1 | 6 | 794 | ||
GenBank mRNA | AF327431.1 | - | 6 | 666 | ||
AK089276.1 | - | 6 | 633 | |||
AK142397.1 | - | 4 | 829 | |||
AK143217.1 | - | 6 | 795 | |||
AK144607.1 | - | 6 | 775 | |||
AK155733.1 | - | 4 | 1162 | |||
AK157340.1 | - | 6 | 766 | |||
AK187471.1 | - | 1 | 57 | |||
BC003901.1 | - | 6 | 739 | 3601310 |