Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290378.1 | NP_001277307.1 | 3 | 514 | ||
NM_001290379.1 | NP_001277308.1 | 3 | 514 | |||
NM_026887.3 | NP_081163.2 | 3 | 514 | |||
GenBank mRNA | AK005223.1 | - | 3 | 378 | ||
AK045558.1 | - | 3 | 515 | |||
AK047724.1 | - | 3 | 391 | |||
AK053941.1 | - | 3 | 387 | |||
AK167690.1 | - | 3 | 389 | |||
BC046964.1 | - | 3 | 371 | 6485320 |