Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148944.4 | NP_683746.1 | 2 | 147 | ||
GenBank mRNA | AF492840.1 | - | 2 | 132 | ||
AK140071.1 | - | 2 | 149 | |||
AY574269.1 | - | 2 | 98 | |||
BC094889.1 | - | 2 | 193 | |||
BC138851.1 | - | 2 | 146 | 8861873 |