Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033223.2 | NP_001028395.1 | 6 | 1407 | ||
NM_001039354.1 | NP_001034443.1 | 6 | 1449 | |||
NM_001284329.1 | NP_001271258.1 | 6 | 2166 | |||
GenBank mRNA | AK160671.1 | - | 6 | 1356 | ||
BC029721.1 | - | 6 | 1355 | |||
BC105965.1 | - | 5 | 1478 |