Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc14a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171010.1 | NP_001164481.1 | 10 | 1498 | ||
NM_001171011.1 | NP_001164482.1 | 9 | 1233 | |||
NM_028122.4 | NP_082398.1 | 9 | 1293 | |||
GenBank mRNA | AF448798.1 | - | 8 | 1090 | ||
AJ420967.1 | - | 9 | 1152 | |||
AK012066.1 | - | 9 | 1143 | |||
AK041979.1 | - | 9 | 1235 | |||
AK153891.1 | - | 10 | 1403 | |||
BC058594.1 | - | 9 | 1180 | 5364683 | ||
BC086673.1 | - | 9 | 1189 | 30646772 | ||
BC100570.1 | - | 9 | 1180 | 30044353 |